Assay Details
| Gene Symbol: AZI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017346753.1 | XP_017202242.1 | 4 - 5 | 571 | 81 | |
| XM_017346754.1 | XP_017202243.1 | 4 - 5 | 571 | 81 | ||
| XM_017346755.1 | XP_017202244.1 | 4 - 5 | 571 | 81 | ||
| XM_017346756.1 | XP_017202245.1 | 4 - 5 | 571 | 81 | ||
More Information
Set Membership: |
|

AZI2
Genomic Map