Assay Details
| Gene Symbol: DAZL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008266045.1 | XP_008264267.1 | 3 - 4 | 524 | 64 | |
| XM_017346734.1 | XP_017202223.1 | 3 - 4 | 595 | 64 | ||
More Information
Set Membership: |
|

DAZL
Genomic Map