Assay Details
| Gene Symbol: LOC100358333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710149.2 | XP_002710195.1 | 3 - 4 | 1493 | 87 | |
| XM_008254950.2 | XP_008253172.1 | 3 - 4 | 1571 | 87 | ||
More Information
Set Membership: |
|

LOC100358333
Genomic Map