Assay Details
| Gene Symbol: RANGAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721407.3 | XP_002721453.1 | 709 | 60 | ||
| XM_008275173.2 | XP_008273395.1 | 994 | 60 | |||
| XM_017350934.1 | XP_017206423.1 | 358 | 60 | |||
More Information
Set Membership: |
|

RANGAP1
Genomic Map