Assay Details
| Gene Symbol: C19H17orf50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017349072.1 | XP_017204561.1 | 1 - 2 | 1339 | 117 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017349072.1 transcripts.
Set Membership: |
|

C19H17orf50
Genomic Map