Assay Details
| Gene Symbol: GAPDHS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722238.2 | XP_002722284.1 | 735 | 59 | ||
| XM_008249497.2 | XP_008247719.1 | 761 | 59 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GAPDHS| Gene Symbol: GAPDHS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722238.2 | XP_002722284.1 | 735 | 59 | ||
| XM_008249497.2 | XP_008247719.1 | 761 | 59 | |||
Set Membership: |
|