Assay Details
| Gene Symbol: LOC100354960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017346745.1 | XP_017202234.1 | 560 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017346745.1 transcripts.
Set Membership: |
|

LOC100354960
Genomic Map