Assay Details
| Gene Symbol: RDH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719562.3 | XP_002719608.2 | 2 - 3 | 536 | 66 | |
| XM_008271832.2 | XP_008270054.1 | 2 - 3 | 536 | 66 | ||
| XM_008271833.2 | XP_008270055.1 | 2 - 3 | 227 | 66 | ||
More Information
Set Membership: |
|

RDH11
Genomic Map