Assay Details
| Gene Symbol: BSPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008252600.2 | XP_008250822.1 | 361 | 65 | ||
| XM_008252601.2 | XP_008250823.1 | 342 | 65 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BSPH1| Gene Symbol: BSPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008252600.2 | XP_008250822.1 | 361 | 65 | ||
| XM_008252601.2 | XP_008250823.1 | 342 | 65 | |||
Set Membership: |
|