Assay Details
| Gene Symbol: LOC100346394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718585.3 | XP_002718631.1 | 7 - 8 | 1336 | 50 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100346394| Gene Symbol: LOC100346394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718585.3 | XP_002718631.1 | 7 - 8 | 1336 | 50 | |
Set Membership: |
|