Assay Details
| Gene Symbol: LOC100345779 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267614.2 | XP_008265836.1 | 12 - 13 | 1997 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100345779| Gene Symbol: LOC100345779 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267614.2 | XP_008265836.1 | 12 - 13 | 1997 | 117 | |
Set Membership: |
|