Assay Details
| Gene Symbol: FAM216A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272215.2 | XP_008270437.1 | 4 - 5 | 471 | 86 | |
| XM_008272216.2 | XP_008270438.1 | 4 - 5 | 465 | 86 | ||
| XM_008272217.2 | XP_008270439.1 | 4 - 5 | 486 | 86 | ||
| XM_008272218.2 | XP_008270440.1 | 4 - 5 | 480 | 86 | ||
More Information
Set Membership: |
|

FAM216A
Genomic Map