Assay Details
| Gene Symbol: C9H3orf84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260643.2 | XP_008258865.1 | 3 - 4 | 446 | 67 | |
| XM_008260646.2 | XP_008258868.1 | 2 - 3 | 304 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008260646.2 transcripts.
Set Membership: |
|

C9H3orf84
Genomic Map