Assay Details
| Gene Symbol: LOC100346409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008275329.2 | XP_008273551.1 | 2503 | 92 | ||
| XM_008275330.2 | XP_008273552.1 | 2312 | 92 | |||
| XM_017337556.1 | XP_017193045.1 | 2503 | 92 | |||
| XM_017337557.1 | XP_017193046.1 | 2044 | 92 | |||
More Information
Set Membership: |
|

LOC100346409
Genomic Map