Assay Details
| Gene Symbol: LOC100346409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008275329.2 | XP_008273551.1 | 581 | 67 | ||
| XM_008275330.2 | XP_008273552.1 | 390 | 67 | |||
| XM_008275331.2 | XP_008273553.1 | 581 | 67 | |||
| XM_017337556.1 | XP_017193045.1 | 581 | 67 | |||
| XM_017337557.1 | XP_017193046.1 | 122 | 67 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008275330.2, XM_017337557.1 transcripts.
Set Membership: |
|

LOC100346409
Genomic Map