Assay Details
| Gene Symbol: DNAAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717733.3 | XP_002717779.1 | 2 - 3 | 318 | 94 | |
| XM_002717734.3 | XP_002717780.1 | 2 - 3 | 318 | 94 | ||
| XM_017348003.1 | XP_017203492.1 | 2 - 3 | 318 | 94 | ||
More Information
Set Membership: |
|

DNAAF4
Genomic Map