Assay Details
| Gene Symbol: CBLIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709184.2 | XP_002709230.1 | 6 - 7 | 874 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CBLIF| Gene Symbol: CBLIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709184.2 | XP_002709230.1 | 6 - 7 | 874 | 62 | |
Set Membership: |
|