Assay Details
| Gene Symbol: LOC100347603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017349788.1 | XP_017205277.1 | 21 - 22 | 3255 | 109 | |
| XM_017349789.1 | XP_017205278.1 | 20 - 21 | 3153 | 109 | ||
| XM_017349790.1 | XP_017205279.1 | 20 - 21 | 3120 | 109 | ||
| XM_017349791.1 | XP_017205280.1 | 20 - 21 | 3099 | 109 | ||
| XM_017349792.1 | XP_017205281.1 | 19 - 20 | 3018 | 109 | ||
| XM_017349793.1 | XP_017205282.1 | 19 - 20 | 2997 | 109 | ||
| XM_017349794.1 | XP_017205283.1 | 19 - 20 | 2964 | 109 | ||
| XM_017349795.1 | XP_017205284.1 | 18 - 19 | 2862 | 109 | ||
More Information
Set Membership: |
|

LOC100347603
Genomic Map