Assay Details
| Gene Symbol: LOC100347603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017349788.1 | XP_017205277.1 | 3 - 4 | 217 | 94 | |
| XM_017349789.1 | XP_017205278.1 | 3 - 4 | 217 | 94 | ||
| XM_017349790.1 | XP_017205279.1 | 3 - 4 | 217 | 94 | ||
| XM_017349791.1 | XP_017205280.1 | 3 - 4 | 217 | 94 | ||
| XM_017349792.1 | XP_017205281.1 | 3 - 4 | 217 | 94 | ||
| XM_017349793.1 | XP_017205282.1 | 3 - 4 | 217 | 94 | ||
| XM_017349794.1 | XP_017205283.1 | 3 - 4 | 217 | 94 | ||
| XM_017349795.1 | XP_017205284.1 | 3 - 4 | 217 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017349788.1, XM_017349789.1, XM_017349790.1, XM_017349791.1, XM_017349792.1, XM_017349793.1, XM_017349794.1, XM_017349795.1 transcripts.
Set Membership: |
|

LOC100347603
Genomic Map