Assay Details
| Gene Symbol: MAMDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708194.3 | XP_002708240.1 | 5 - 6 | 1452 | 60 | |
| XM_017341729.1 | XP_017197218.1 | 5 - 6 | 1460 | 60 | ||
More Information
Set Membership: |
|

MAMDC2
Genomic Map