Assay Details
| Gene Symbol: BRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720786.3 | XP_002720832.1 | 636 | 56 | ||
| XM_017350369.1 | XP_017205858.1 | 577 | 56 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BRF2| Gene Symbol: BRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720786.3 | XP_002720832.1 | 636 | 56 | ||
| XM_017350369.1 | XP_017205858.1 | 577 | 56 | |||
Set Membership: |
|