Assay Details
| Gene Symbol: PLEKHG4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008251397.2 | XP_008249619.1 | 1814 | 62 | ||
| XM_017339228.1 | XP_017194717.1 | 1814 | 62 | |||
| XM_017339229.1 | XP_017194718.1 | 1814 | 62 | |||
More Information
Set Membership: |
|

PLEKHG4B
Genomic Map