Assay Details
| Gene Symbol: WIPF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712298.3 | XP_002712344.1 | 4 - 5 | 587 | 67 | |
| XM_008258766.2 | XP_008256988.1 | 4 - 5 | 541 | 67 | ||
| XM_008258767.2 | XP_008256989.1 | 4 - 5 | 466 | 67 | ||
More Information
Set Membership: |
|

WIPF1
Genomic Map