Assay Details
Gene Symbol: CCDC191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002716832.3 | XP_002716878.1 | 14 - 15 | 2431 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CCDC191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002716832.3 | XP_002716878.1 | 14 - 15 | 2431 | 62 |
Set Membership: |
![]() ![]() ![]() ![]() |