Assay Details
Gene Symbol: TSGA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017342679.1 | XP_017198168.1 | 7 - 8 | 907 | 85 | |
XM_017342680.1 | XP_017198169.1 | 7 - 8 | 904 | 85 | ||
XM_017342681.1 | XP_017198170.1 | 6 - 7 | 733 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
