Assay Details
| Gene Symbol: LOC103349065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260021.2 | XP_008258243.1 | 1 | 624 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008260021.2 transcripts.
Set Membership: |
|

LOC103349065
Genomic Map