Assay Details
| Gene Symbol: PPP1R42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710453.3 | XP_002710499.1 | 5 - 6 | 774 | 77 | |
| XM_017341366.1 | XP_017196855.1 | 5 - 6 | 769 | 77 | ||
| XM_017341367.1 | XP_017196856.1 | 5 - 6 | 737 | 77 | ||
| XM_017341368.1 | XP_017196857.1 | 5 - 6 | 770 | 77 | ||
More Information
Set Membership: |
|

PPP1R42
Genomic Map