Assay Details
| Gene Symbol: RAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717880.3 | XP_002717926.1 | 1 - 2 | 215 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002717880.3 transcripts.
Set Membership: |
|

RAM1
Genomic Map