Assay Details
| Gene Symbol: PIGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008254442.2 | XP_008252664.1 | 6 - 7 | 781 | 89 | |
| XM_008254443.2 | XP_008252665.1 | 5 - 6 | 685 | 89 | ||
| XM_008254444.2 | XP_008252666.1 | 5 - 6 | 684 | 89 | ||
More Information
Set Membership: |
|

PIGF
Genomic Map