Assay Details
| Gene Symbol: PFN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710063.3 | XP_002710109.1 | 2 - 3 | 490 | 69 | |
| XM_008254800.2 | XP_008253022.1 | 2 - 3 | 376 | 69 | ||
More Information
Set Membership: |
|

PFN4
Genomic Map