Assay Details
| Gene Symbol: ZSCAN21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008248904.2 | XP_008247126.1 | 598 | 89 | ||
| XM_017337859.1 | XP_017193348.1 | 654 | 89 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZSCAN21| Gene Symbol: ZSCAN21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008248904.2 | XP_008247126.1 | 598 | 89 | ||
| XM_017337859.1 | XP_017193348.1 | 654 | 89 | |||
Set Membership: |
|