Assay Details
| Gene Symbol: ZSCAN21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008248904.2 | XP_008247126.1 | 771 | 108 | ||
| XM_017337859.1 | XP_017193348.1 | 827 | 108 | |||
More Information
Set Membership: |
|

ZSCAN21
Genomic Map