Assay Details
| Gene Symbol: NKIRAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716213.3 | XP_002716259.1 | 3 - 4 | 537 | 98 | |
| XM_008266059.2 | XP_008264281.1 | 3 - 4 | 490 | 98 | ||
| XM_008266060.2 | XP_008264282.1 | 3 - 4 | 447 | 98 | ||
| XM_017346746.1 | XP_017202235.1 | 2 - 3 | 399 | 98 | ||
More Information
Set Membership: |
|

NKIRAS1
Genomic Map