Assay Details
| Gene Symbol: PSMB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719331.3 | XP_002719377.1 | 5 - 6 | 668 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PSMB3| Gene Symbol: PSMB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719331.3 | XP_002719377.1 | 5 - 6 | 668 | 61 | |
Set Membership: |
|