Assay Details
| Gene Symbol: UFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008252119.2 | XP_008250341.1 | 139 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008252119.2 transcripts.
Set Membership: |
|

UFD1
Genomic Map