Assay Details
| Gene Symbol: UFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723570.3 | XP_002723616.1 | 519 | 57 | ||
| XM_008252119.2 | XP_008250341.1 | 262 | 57 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UFD1| Gene Symbol: UFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723570.3 | XP_002723616.1 | 519 | 57 | ||
| XM_008252119.2 | XP_008250341.1 | 262 | 57 | |||
Set Membership: |
|