Assay Details
| Gene Symbol: EPM2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714860.3 | XP_002714906.1 | 2 - 3 | 2846 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPM2A| Gene Symbol: EPM2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714860.3 | XP_002714906.1 | 2 - 3 | 2846 | 60 | |
Set Membership: |
|