Assay Details
| Gene Symbol: FZD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710725.3 | XP_002710771.1 | 3 - 4 | 755 | 60 | |
| XM_017341467.1 | XP_017196956.1 | 3 - 4 | 730 | 60 | ||
More Information
Set Membership: |
|

FZD6
Genomic Map