Assay Details
| Gene Symbol: MTHFS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722962.3 | XP_002723008.1 | 239 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MTHFS| Gene Symbol: MTHFS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722962.3 | XP_002723008.1 | 239 | 81 | ||
Set Membership: |
|