Assay Details
| Gene Symbol: PTPN11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272268.2 | XP_008270490.1 | 5 - 6 | 890 | 56 | |
| XM_008272269.2 | XP_008270491.1 | 5 - 6 | 892 | 56 | ||
| XM_008272270.2 | XP_008270492.1 | 5 - 6 | 886 | 56 | ||
More Information
Set Membership: |
|

PTPN11
Genomic Map