Assay Details
| Gene Symbol: GPR162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712902.3 | XP_002712948.1 | 4 - 5 | 1755 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR162| Gene Symbol: GPR162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712902.3 | XP_002712948.1 | 4 - 5 | 1755 | 93 | |
Set Membership: |
|