Assay Details
| Gene Symbol: ARHGAP24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267631.2 | XP_008265853.1 | 6 - 7 | 1109 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARHGAP24| Gene Symbol: ARHGAP24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267631.2 | XP_008265853.1 | 6 - 7 | 1109 | 60 | |
Set Membership: |
|