Assay Details
| Gene Symbol: B3GNT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722309.3 | XP_002722355.1 | 354 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002722309.3 transcripts.
Set Membership: |
|

B3GNT8
Genomic Map