Assay Details
| Gene Symbol: TAMM41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713047.3 | XP_002713093.1 | 6 - 7 | 1111 | 91 | |
| XM_017343742.1 | XP_017199231.1 | 6 - 7 | 1111 | 91 | ||
| XM_017343743.1 | XP_017199232.1 | 5 - 6 | 961 | 91 | ||
| XM_017343744.1 | XP_017199233.1 | 5 - 6 | 769 | 91 | ||
| XM_017343745.1 | XP_017199234.1 | 6 - 7 | 825 | 91 | ||
More Information
Set Membership: |
|

TAMM41
Genomic Map