Assay Details
| Gene Symbol: RIOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716749.2 | XP_002716795.1 | 2 - 3 | 533 | 85 | |
| XM_008267074.1 | XP_008265296.1 | 2 - 3 | 533 | 85 | ||
| XM_008267075.1 | XP_008265297.1 | 2 - 3 | 533 | 85 | ||
| XM_008267076.1 | XP_008265298.1 | 2 - 3 | 533 | 85 | ||
More Information
Set Membership: |
|

RIOX2
Genomic Map