Assay Details
| Gene Symbol: RIOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716749.2 | XP_002716795.1 | 7 - 8 | 1158 | 68 | |
| XM_008267076.1 | XP_008265298.1 | 7 - 8 | 1155 | 68 | ||
More Information
Set Membership: |
|

RIOX2
Genomic Map