Assay Details
| Gene Symbol: RIOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716749.2 | XP_002716795.1 | 8 - 9 | 1257 | 101 | |
| XM_008267074.1 | XP_008265296.1 | 8 - 9 | 1266 | 101 | ||
| XM_008267075.1 | XP_008265297.1 | 8 - 9 | 1263 | 101 | ||
| XM_008267076.1 | XP_008265298.1 | 8 - 9 | 1254 | 101 | ||
More Information
Set Membership: |
|

RIOX2
Genomic Map