Assay Details
| Gene Symbol: CDC37L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008255227.2 | XP_008253449.1 | 1 - 2 | 264 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDC37L1| Gene Symbol: CDC37L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008255227.2 | XP_008253449.1 | 1 - 2 | 264 | 88 | |
Set Membership: |
|