Assay Details
| Gene Symbol: NSRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718905.3 | XP_002718951.1 | 3 - 4 | 244 | 67 | |
| XM_017349125.1 | XP_017204614.1 | 4 - 5 | 392 | 67 | ||
More Information
Set Membership: |
|

NSRP1
Genomic Map