Assay Details
| Gene Symbol: LOC100344835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715453.3 | XP_002715499.1 | 9 - 10 | 1356 | 69 | |
| XM_008264782.2 | XP_008263004.1 | 10 - 11 | 1407 | 69 | ||
More Information
Set Membership: |
|

LOC100344835
Genomic Map